missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAIM Protein

A cDNA sequence encoding the FAIM was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11841-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978548

  • 3401.86€ / 1mg


Estimated Shipment
06-04-2021
Add to basket

Description

Description

Specifications

Specifications

FAIM Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FAIM protein solution (0.5 mg /ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLLPFIR TLPLLCYNHL LVSPDSATLS PPYSLEKMTD LVAVWDVALS DGVHKIEFEH GTTSGKRVVY VDGKEEIRKE WMFKLVGKET FYVGAAKTKA TINIDAISGF AYEYTLEING KSLKKYMEDR SKTTNTWVLH MDGENFRIVL EKDAMDVWCN GKKLETAGEF VDDGTETHFS IGNHDCYIKA VSSGKRKEGI IHTLIVDNRE IPEIAS
Videos
SDS
Documents