missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAM3C Protein
A cDNA sequence encoding the FAM3C was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | FAM3C Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948568
|
enQuireBio™
QP11846-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928568
|
enQuireBio™
QP11846-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938568
|
enQuireBio™
QP11846-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
FAM3C Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FAM3C solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 200mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQVFEIKMDA SLGNLFARSA LDTAARSTKP PRYKCGISKA CPEKHFAFKM ASGAANVVGP KICLEDNVLM SGVKNNVGRG INVALANGKT GEVLDTKYFD MWGGDVAPFI EFLKAIQDGT IVLMGTYDDG ATKLNDEARR LIADLGSTSI TNLGFRDNWV FCGGKGIKTK SPFEQHIKNN KDTNKYEGWP EVVEMEGCIP QKQD | |
Greater than 90% as determined by SDS-PAGE. |