missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF 18 Protein

A cDNA sequence encoding the FGF 18 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11869-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978638

  • 212.00€ / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FGF 18 Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASEENVDFRIHV ENQTRARDDV SRKQLRLYQL YSRTSGKHIQ VLGRRISARG EDGDKYAQLL VETDTFGSQV RIKGKETEFY LCMNRKGKLV GKPDGTSKEC VFIEKVLENN YTALMSAKYS GWYVGFTKKG RPRKGPKTRE NQQDVHFMKR YPKGQPELQK PFKYTTVTKR SRRIRPTHPA
Purity as determined by densitometric image analysis is greater than 95%.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FGF18 was filtered (0.4microm) and lyophilized in phosphate buffered saline and 5% w/v trehalose.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions