missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR3 / CD333 Protein
A DNA sequence encoding the Homo sapiens (Human) FGFR3 / CD333, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6046-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000133.1 | |
FGFR3 / CD333 Protein | |
Research Use Only | |
FGFR3 | |
Recombinant Protein | |
E. coli | |
ESLGTEQRVV GRAAEVPGPE PGQQEQLVFG SGDAVELSCP PPGGGPMGPT VWVKDGTGLV PSERVLVGPQ RLQVLNASHE DSGAYSCRQR LTQRVLCHFS VRVTDAPSSG DDEDGEDEAE DTGVDTGAPY WTRPERMDKK LLAVPAANTV RFRCPAAGNP TPSISWLKNG REFRGEHRIG GIKLRHQQWS LVMESVVPSD RGNYTCVVEN KFGSIRQTYT LDVLERSPHR PILQAGLPAN QTAVLGSDVE FHCKVYSDAQ PHIQWLKHVE VNGSKVGPDG TPYVTVLKTA GANTTDKELE VLSLHNVTFE DAGEYTCLAG NSIGFSHHSA WLVVLPAEEE LVEADEAGSV YAG | |
Greater than 90% as determined by SDS-PAGE. |
2261 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |