missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FHL2 Protein

A cDNA sequence encoding the FHL2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11879-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978668

  • 5450.00€ / 1mg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FHL2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTERFDC HHCNESLFGK KYILREESPY CVVCFETLFA NTCEECGKPI GCDCKDLSYK DRHWHEACFH CSQCRNSLVD KPFAAKEDQL LCTDCYSNEY SSKCQECKKT IMPGTRKMEY KGSSWHETCF ICHRCQQPIG TKSFIPKDNQ NFCVPCYEKQ HAMQCVQCKK PITTGGVTYR EQPWHKECFV CTACRKQLSG QRFTARDDFA YCLNCFCDLY AKKCAGCTNP ISGLGGTKYI SFEERQWHND CFNCKKCSLS LVGRGFLTER DDILCPDCGK DI
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FHL2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions