missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroblast growth factor 13 Protein

A DNA sequence encoding the Homo sapiens (Human) Fibroblast growth factor 13, was expressed in the hosts and tags indicated.

160.00€ - 2385.00€

Specifications

Accession Number NP_001132970.1
Gene ID (Entrez) 2258
Name Fibroblast growth factor 13 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15979549
View Documents Promotion Details
enQuireBio™
QP6042-EC-100UG
100 μg
530.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15999549
View Documents Promotion Details
enQuireBio™
QP6042-EC-1MG
1 mg
2385.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15989549
View Documents Promotion Details
enQuireBio™
QP6042-EC-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15909559
View Documents Promotion Details
enQuireBio™
QP6042-EC-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15919559
View Documents Promotion Details
enQuireBio™
QP6042-EC-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15929559
View Documents Promotion Details
enQuireBio™
QP6042-EC-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001132970.1
Fibroblast growth factor 13 Protein
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
2258
Research Use Only
FGF13
Recombinant Protein
E. coli
MAAAIASSLI RQKRQARERE KSNACKCVSS PSKGKTSCDK NKLNVFSRVK LFGSKKRRRR RPEPQLKGIV TKLYSRQGYH LQLQADGTID GTKDEDSTYT LFNLIPVGLR VVAIQGVQTK LYLAMNSEGY LYTSELFTPE CKFKESVFEN YYVTYSSMIY RQQQSGRGWY LGLNKEGEIM KGNHVKKNKP AAHFLPKPLK VAMYKEPSLH DLTEFSRSGS GTPTKSRSVS GVLNGGKSMS HNEST
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications