missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fibulin 5 Protein

Product Code. 18747247
Click to view available options
:
50μg
This item is not returnable. View return policy

Product Code. 18747247

Brand: R&D Systems™ 9006FB050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Fibulin 5 Protein is derived from HEK293. The Recombinant Human Fibulin 5 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UBX5
For Use With (Application) Bioactivity
Gene ID (Entrez) 10516
Molecular Weight (g/mol) Observed M.W.: 60-72kDa, reducing conditions, Theoretical M.W.: 49kDa
Quantity 50μg
Source Human embryonic kidney cell,HEK293-derived human Fibulin 5/DANCE protein Gln24-Phe448,with an N-terminal HA tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, FBLN5, fibulin 5, UP50, UP50FLJ90059, Urine p50 protein
Common Name Fibulin 5/DANCE
Gene Symbol FBLN5
Biological Activity Measured by the ability of the immobilized protein to enhance the adhesion of HUVEC human umbilical vein endothelial cells. The ED50 for this effect is typically 15-90ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Gln24-Phe448, with an N-terminal HA tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.