missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FIS1 Protein

A cDNA sequence encoding the FIS1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11884-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948688

  • 212.00€ / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

FIS1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEAVLNELVS VEDLLKFEKK FQSEKAAGSV SKSTQFEYAW CLVRSKYNDD IRKGIVLLEE LLPKGSKEEQ RDYVFYLAVG NYRLKEYEKA LKYVRGLLQT EPQNNQAKEL ERLIDKAMKK DG
Greater than 95% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FIS1 protein solution (1 mg/ml) containing 20mM Tris-HCl pH 8 and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions