missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FKBP6 Protein

Product Code. p-7148414
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15934408

Brand: enQuireBio™ QP106102ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Gene ID (Entrez) 8468
Name FKBP6 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol FKBP6
Biological Activity Specific activity is > 190 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Product Type Recombinant Protein
Cross Reactivity Human
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGGSALNQGV LEGDDAPGQS LYERLSQRML DISGDRGVLK DVIREGAGDL VAPDASVLVK YSGYLEHMDR PFDSNYFRKT PRLMKLGEDI TLWGMELGLL SMRRGELARF LFKPNYAYGT LGCPPLIPPN TTVLFEIELL DFLDCAESDK FCALSAEQQD QFPLQKVLKV AATEREFGNY LFRQNRFYDA KVRYKRALLL LRRRSAPPEE QHLVEAAKLP VLLNLSFTYL KLDRPTIALC YGEQALIIDQ KNAKALFRCG QACLLLTEYQ KARDFLVRAQ KEQPFNHDIN NELKKLASCY RDYVDKEKEM WHRMFAPCGD GSTAGES
Buffer FKBP6 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, 1mM EDTA, 50mM NaCl and 40% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.