missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FKBP6 Protein
Recombinant Protein
Brand: enQuireBio™ QP10610-1mg
Additional Details : Weight : 0.01000kg
Specifications
8468 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
FKBP6 | |
Recombinant Protein | |
His | |
FKBP6 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT, 1mM EDTA, 50mM NaCl and 40% glycerol. |
FKBP6 Protein | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 190 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
MGSSHHHHHH SSGLVPRGSH MGGSALNQGV LEGDDAPGQS LYERLSQRML DISGDRGVLK DVIREGAGDL VAPDASVLVK YSGYLEHMDR PFDSNYFRKT PRLMKLGEDI TLWGMELGLL SMRRGELARF LFKPNYAYGT LGCPPLIPPN TTVLFEIELL DFLDCAESDK FCALSAEQQD QFPLQKVLKV AATEREFGNY LFRQNRFYDA KVRYKRALLL LRRRSAPPEE QHLVEAAKLP VLLNLSFTYL KLDRPTIALC YGEQALIIDQ KNAKALFRCG QACLLLTEYQ KARDFLVRAQ KEQPFNHDIN NELKKLASCY RDYVDKEKEM WHRMFAPCGD GSTAGES |