missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FLT1 D4 Protein

A cDNA sequence encoding the FLT1 D4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10613-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15924418

  • 125.00€ / 2µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

2321
Greater than 90.0% as determined by(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE.
Research Use Only
FLT1
Recombinant Protein
Insect
MVSYWDTGVL LCALLSCLLL TGSSSGSKLK DPELSLKGTQ HIMQAGQTLH LQCRGEAAHK WSLPEMVSKE SERLSITKSA CGRNGKQFCS TLTLNTAQAN HTGFYSCKYL AVPTSKKKET ESAIYIFISD TGRPFVEMYS EIPEIIHMTE GRELVIPCRV TSPNITVTLK KFPLDTLIPD GKRIIWDSRK GFIISNATYK EIGLLTCEAT VNGHLYKTNY LTHRQTNTII DVQISTPRPV KLLRGHTLVL NCTATTPLNT RVQMTWSYPD EKNKRASVRR RIDQSNSHAN IFYSVLTIDK MQNKDKGLYT CRVRSGPSFK SVNTSVHIYD KAFITVKHRK QQVLETVAGK RSYRLSMKVK AFPSPEVVWL KDGLPATEKS ARYLTRGYSL IIKDVTEEDA GNYTILLSIK QSNVFKNLTA TLIVNVKPQI YEKAVSSFPD PALYPLGSRQ ILTCTAYGIP QPTIKWFWHP CNHNHSEARC DFCSNNEESF ILDADSNMGN RIESITQRMA IIEGKNKMAS TLVVADSRIS GIYICIASNK VGTVGRNISF YITDVPNGFH VNLEKMPTEG EDLKLSCTVN KFLYRDVTWI LLRTVNNRTM HYSISKQKMA ITKEHSITLN LTIMNVSLQD SGTYACRARN VYTGEEILQK KEITIRDQEA PYLLRNLSDH TVAISSSTTL DCHANGVPEP QITWFKNNHK IQQEPGIILG PGSSTLFIER VTEEDEGVYH CKATNQKGSV ESSAYLTVQG TSDKSNLELI TLTCTCVAAT LFWLLLTLFI RKMKRSSSEI KTDYLSIIMD PDEVPLDEQC ERLPYDASKW EFARERLKLG KSLGRGAFGK VVQASAFGIK KSPTCRTVAV KMLKEGATAS EYKALMTELK ILTHIGHHLN VVNLLGACTK QGGPLMVIVE YCKYGNLSNY LKSKRDLFFL NKDAALHMEP KKEKMEPGLE QGKKPRLDSV TSSESFASSG FQEDKSLSDV EEEEDSDGFY KEPITMEDLI SYSFQVARGM EFLSSRKCIH RDLAARNILL SENNVVKICD FGLARDIYKN PDYVRKGDTR LPLKWMAPES IFDKIYSTKS DVWSYGVLLW EIFSLGGSPY PGVQMDEDFC SRLREGMRMR APEYSTPEIY QIMLDCWHRD PKERPRFAEL VEKLGDLLQA NVQQDGKDYI PINAILTGNS GFTYSTPAFS EDFFKESISA PKFNSGSSDD VRYVNAFKFM SLERIKTFEE LLPNATSMFD DYQGDSSTLL ASPMLKRFTW TDSKPKASLK IDLRVTSKSK ESGLSDVSRP SFCHSSCGHV SEGKRRFTYD HAELERKIAC CSPPPDYNSV VLYSTPPI
FLT1 D4 Protein
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The activity of FLT1D1-4 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces, and in Far-Western and cross-linking experiments with iodinated VEGF.
Human
Untagged
FLT1 D1-4 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions