missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FOS-B Protein
A cDNA sequence encoding the FOS-B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11898-10ug
Additional Details : Weight : 0.01000kg
Specifications
2354 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MFQAFPGDYD SGSRCSSSPS AESQYLSSVD SFGSPPTAAA SQECAGLGEM PGSFVPTVTA ITTSQDLQWL VQPTLISSMA QSQGQPLASQ PPVVDPYDMP GTSYSTPGMS GYSSGGASGS GGPSTSGTTS GPGPARPARA RPRRPREETL TPEEEEKRRV RRERNKLAAA KCRNRRRELT DRLQAETDQL EEEKAELESE IAELQKEKER LEFVLVAHKP GCKIPYEEGP GPGPLAEVRD LPGSAPAKED GFSWLLPPPP PPPLPFQTSQ DAPPNLTASL FTHSEVQVLG DPFPVVNPSY TSSFVLTCPE VSAFAGAQRT SGSDQPSDPL NSPSLLAL |
FOS-B Protein | |
Research Use Only | |
FOSB | |
Human | |
Untagged | |
FOS-B in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |