missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein

Recombinant Protein

2600.00€ - 4240.00€

Specifications

Accession Number NP_001291321.1
Gene ID (Entrez) 6376
Name Fractalkine / CX3CL1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15925879
View Documents Promotion Details
enQuireBio™
QP5299-500UG
500 μg
2600.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915879
View Documents Promotion Details
enQuireBio™
QP5299-1MG
1 mg
4240.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001291321.1
Fractalkine / CX3CL1 Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human Fractalkine / CX3CL1 induces Human PBMC chemotaxis (significanty different from control at Human Fractalkine / CX3CL1 concentrations greater than 10-100 ng/ml).
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
6376
Research Use Only
CX3CL1
Recombinant Protein
E. coli
QHHGVTKCNI . TCSKMTSKI P. VALLIHYQ QN. QASCGKR AII. LETRQH RLFC. ADPKE QWVKD. AMQH LDRQAA. ALT RNG
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications