missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FSH Protein

A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name FSH Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15934438
View Documents Promotion Details
enQuireBio™
QP10620-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15914438
View Documents Promotion Details
enQuireBio™
QP10620-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15924438
View Documents Promotion Details
enQuireBio™
QP10620-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

FSH Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
Human (HEK293)
FSH subunit alpha: APDVQDCPECTLQENPFFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKSFSH subunit beta: NSCELTNITIAIEKEECRFCISINTTWCAGYCYTRDLVYKDPARPKIQKTCTFKELVYETVRVPGCAHHADSLYTYPVATQCHCGKCDSDSTDCTVRGLGPSYCSFGEMKE
Greater than 95% as determined by SDS-PAGE.
Research Use Only
The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml.
Human
Untagged
The recombinant FSH was lyophilized from a concentrated (1 mg/ml) solution containing PBS.
Videos
SDS
Documents

Documents

Product Certifications