missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fusion glycoprotein F0 Protein

Product Code. p-7155766
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16058541

Brand: enQuireBio™ QP9634ye10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Fusion glycoprotein F0, was expressed in the hosts and tags indicated.

Specifications

Name Fusion glycoprotein F0 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol F
Protein Length Extracellular Domain
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence NITEEFYQST CSAVSKGYLS ALRTGWYTSV ITIELSNIKE NKCNGTDAKV KLIKQELDKY KNAVTELQLL MQSTPPTNNR ARRELPRFMN YTLNNAKKTN VTLSKKRKRR FLGFLLGVGS AIASGVAVSK VLHLEGEVNK IKSALLSTNK AVVSLSNGVS VLTSKVLDLK NYIDKQLLPI VNKQSCSISN IETVIEFQQK NNRLLEITRE FSVNAGVTTP VSTYMLTNSE LLSLINDMPI TNDQKKLMSN NVQIVRQQSY SIMSIIKEEV LAYVVQLPLY GVIDTPCWKL HTSPLCTTNT KEGSNICLTR TDRGWYCDNA GSVSFFPQAE TCKVQSNRVF CDTMNSLTLP SEINLCNVDI FNPKYDCKIM TSKTDVSSSV ITSLGAIVSC YGKTKCTASN KNRGIIKTFS NGCDYVSNKG MDTVSVGNTL YYVNKQEGKS LYVKGEPIIN FYDPLVFPSD EFDASISQVN EKINQSLAFI RKSDELLHNV NAGKSTTNIM ITT
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.