missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GARS Protein

A cDNA sequence encoding the GARS was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name GARS Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15998838
View Documents Promotion Details
enQuireBio™
QP11933-HIS-INSECT-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15978838
View Documents Promotion Details
enQuireBio™
QP11933-HIS-INSECT-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15988838
View Documents Promotion Details
enQuireBio™
QP11933-HIS-INSECT-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

GARS Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GARS is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol.
Research Use Only
Recombinant Protein
Baculovirus
MPSPRPVLLR GARAALLLLL PPRLLARPSL LLRRSLSAAS CPPISLPAAA SRSSMDGAGA EEVLAPLRLA VRQQGDLVRK LKEDKAPQVD VDKAVAELKA RKRVLEAKEL ALQPKDDIVD RAKMEDTLKR RFFYDQAFAI YGGVSGLYDF GPVGCALKNN IIQTWRQHFI QEEQILEIDC TMLTPEPVLK TSGHVDKFAD FMVKDVKNGE CFRADHLLKA HLQKLMSDKK CSVEKKSEME SVLAQLDNYG QQELADLFVN YNVKSPITGN DLSPPVSFNL MFKTFIGPGG NMPGYLRPET AQGIFLNFKR LLEFNQGKLP FAAAQIGNSF RNEISPRSGL IRVREFTMAE IEHFVDPSEK DHPKFQNVAD LHLYLYSAKA QVSGQSARKM RLGDAVEQGV INNTVLGYFI GRIYLYLTKV GISPDKLRFR QHMENEMAHY ACDCWDAESK TSYGWIEIVG CADRSCYDLS CHARATKVPL VAEKPLKEPK TVNVVQFEPS KGAIGKAYKK DAKLVMEYLA ICDECYITEM EMLLNEKGEF TIETEGKTFQ LTKDMINVKR FQKTLYVEEV VPNVIEPSFG LGRIMYTVFE HTFHVREGDE QRTFFSFPAV VAPFKCSVLP LSQNQEFMPF VKELSEALTR HGVSHKVDDS SGSIGRRYAR TDEIGVAFGV TIDFDTVNKT PHTATLRDRD SMRQIRAEIS ELPSIVQDLA NGNITWADVE ARYPLFEGQE TGKKETIEE
Greater than 80.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications