missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GAS7 Protein

A cDNA sequence encoding the GAS7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11934-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928848

  • 189.00€ / 50µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

GAS7 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GAS7 protein solution contains 20mM Tris-HCl, pH 8.0, 100mM NaCl, 2mM DTT and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MKPGMVPPPP GEESQTVILP PGWQSYLSPQ GRRYYVNTTT NETTWERPSS SPGIPASPGS HRSSLPPTVN GYHASGTPAH PPETAHMSVR KSTGDSQNLG SSSPSKKQSK ENTITINCVT FPHPDTMPEQ QLLKPTEWSY CDYFWADKKD PQGNGTVAGF ELLLQKQLKG KQMQKEMSEF IRERIKIEED YAKNLAKLSQ NSLASQEEGS LGEAWAQVKK SLADEAEVHL KFSAKLHSEV EKPLMNFREN FKKDMKKCDH HIADLRKQLA SRYASVEKAR KALTERQRDL EMKTQQLEIK LSNKTEEDIK KARRKSTQAG DDLMRCVDLY NQAQSKWFEE MVTTTLELER LEVERVEMIR QHLCQYTQLR HETDMFNQST VEPVDQLLRK VDPAKDRELW VREHKTGNIR PVDMEI
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions