missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GDF-15, Animal-Free Protein

Product Code. 18755975
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
This item is not returnable. View return policy

Product Code. 18755975

Brand: R&D Systems™ Qk0170500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Accession Number Q99988.3
Formulation Lyophilized from acetonitrile/TFA
Gene ID (Entrez) 9518
Molecular Weight (g/mol) M.W. (Observed): Dimeric GDF-15 protein only, M.W. (Theroretical): 25 kDa (dimer)
Quantity 500 μg
Source E. coli-derived human GDF-15 protein
Storage Requirements Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Conjugate Unconjugated
Reconstitution Resuspend in 10 mM HCl at >100 μg/mL, prepare single use aliquots, add carrier protein if desired
Protein GDF-15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.