missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF-8 / Myostatin / MSTN Protein

Product Code. p-7152059
Click to view available options
Quantity:
10 μg
2 μg
Unit Size:
10µg
2µg
This item is not returnable. View return policy

Product Code. 15925919

Brand: enQuireBio™ QP53292ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_005250.1
Gene ID (Entrez) 2660
Name GDF-8 / Myostatin / MSTN Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol MSTN
Biological Activity MPC-11 cell cytotoxicity significantly induced by less than 50 ng/ml which corresponds to an activity of at least 2.0 x 10^4 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS
Buffer Sterile filtered (0.22 um) solution containing 0.1% TFA
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.