missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GDF10 Protein
A cDNA sequence encoding the GDF10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11947-1mg
Additional Details : Weight : 0.01000kg
Specifications
2662 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MQWDEPRVCS RRYLKVDFAD IGWNEWIISP KSFDAYYCAG ACEFPMPKIV RPSNHATIQS IVRAVGIIPG IPEPCCVPDK MNSLGVLFLD ENRNVVLKVY PNMSVDTCAC R | |
Greater than 95.0% as determined by SDS-PAGE. |
GDF10 Protein | |
Research Use Only | |
GDF10 | |
Human | |
Untagged | |
The GDF10 solution (0.5 mg/ml) contains 10mM Sodium citrate (pH 3.5), 1mM DTT, 40% glycerol and 0.1M NaCl. |