missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GIPC2 Protein

A cDNA sequence encoding the GIPC2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11976-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978978

  • 3401.86€ / 1mg


Estimated Shipment
26-03-2021
Add to basket

Description

Description

Specifications

Specifications

GIPC2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GIPC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 30% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPLKLR GKKKAKSKET AGLVEGEPTG AGGGSLSASR APARRLVFHA QLAHGSATGR VEGFSSIQEL YAQIAGAFEI SPSEILYCTL NTPKIDMERL LGGQLGLEDF IFAHVKGIEK EVNVYKSEDS LGLTITDNGV GYAFIKRIKD GGVIDSVKTI CVGDHIESIN GENIVGWRHY DVAKKLKELK KEELFTMKLI EPKKAFEIEP RSKAGKSSGE KIGCGRATLR LRSKGPATVE EMPSETKAKA IEKIDDVLEL YMGIRDIDLA TTMFEAGKDK VNPDEFAVAL DETLGDFAFP DEFVFDVWGV IGDAKRRGL
Videos
SDS
Documents