Learn More
enQuireBio™ Recombinant Human GKN3P Protein
A cDNA sequence encoding the GKN3P was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11979-10ug
Additional Details : Weight : 0.01000kg
Specifications
GKN3P Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASMMNIRFNHPL YGSFGTQIIH IGAFQGMVSI RDNNIFSEWD GILDYKNALL VAKVFNKMAC VLARMDKAVF PSLDDISKAL DKQAFKYYPS TRGLTYTVLP SWVKNLAQYG KPIKNMCRDD PTYFAQQQKE GTALAIDSNS CFEIQLLSFM GLFICGETPG L | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GKN3P filtered (0.4microm) solution at a concentration of 0.3 mg/ml in 0.03M acetate buffer, pH 4.0 and 20% glycerol. |