missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GLIPR2 Protein

A cDNA sequence encoding the GLIPR2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11986-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979008

  • 212.00€ / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GLIPR2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGKSASKQFH NEVLKAHNEY RQKHGVPPLK LCKNLNREAQ QYSEALASTR ILKHSPESSR GQCGENLAWA SYDQTGKEVA DRWYSEIKNY NFQQPGFTSG TGHFTAMVWK NTKKMGVGKA SASDGSSFVV ARYFPAGNVV NEGFFEENVL PPKK
Greater than 90.0% as determined by SDS-PAGE.
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GLIPR2 1 mg/ml protein contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M DTT and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions