missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLTPD1 Protein
A cDNA sequence encoding the GLTPD1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 1815.00€
Specifications
Gene ID (Entrez) | 80772 |
---|---|
Name | GLTPD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CPTP |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929038
|
enQuireBio™
QP11994-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919038
|
enQuireBio™
QP11994-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909038
|
enQuireBio™
QP11994-100UG |
100 μg |
1815.00€
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
80772 | |
Research Use Only | |
CPTP | |
Human | |
His | |
GLTPD1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol and 1mM DTT. |
GLTPD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDSETG FNLKVVLVSF KQCLDEKEEV LLDPYIASWK GLVRFLNSLG TIFSFISKDV VSKLRIMERL RGGPQSEHYR SLQAMVAHEL SNRLVDLERR SHHPESGCRT VLRLHRALHW LQLFLEGLRT SPEDARTSAL CADSYNASLA AYHPWVVRRA VTVAFCTLPT REVFLEAMNV GPPEQAVQML GEALPFIQRV YNVSQKLYAE HSLLDLP | |
Greater than 90.0% as determined by SDS-PAGE. |