missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GOLGA7 Protein

A cDNA sequence encoding the GOLGA7 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name GOLGA7 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15929128
View Documents Promotion Details
enQuireBio™
QP12024-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15919128
View Documents Promotion Details
enQuireBio™
QP12024-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15909128
View Documents Promotion Details
enQuireBio™
QP12024-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

GOLGA7 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
GOLGA7 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMRPQQAP VSGKVFIQRD YSSGTRCQFQ TKFPAELENR IDRQQFEETV RTLNNLYAEA EKLGGQSYLE GCLACLTAYT IFLCMETHYE KVLKKVSKYI QEQNEKIYAP QGLLLTDPIE RGLRVIEITI YEDRGMSSGR
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications