missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GOLGA7 Protein
A cDNA sequence encoding the GOLGA7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12024-5ug
Additional Details : Weight : 0.01000kg
Specifications
GOLGA7 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GOLGA7 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRPQQAP VSGKVFIQRD YSSGTRCQFQ TKFPAELENR IDRQQFEETV RTLNNLYAEA EKLGGQSYLE GCLACLTAYT IFLCMETHYE KVLKKVSKYI QEQNEKIYAP QGLLLTDPIE RGLRVIEITI YEDRGMSSGR |