missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GREM2 Protein

A cDNA sequence encoding the GREM2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12049-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979198

  • 3401.86€ / 1mg


Estimated Shipment
01-04-2021
Add to basket

Description

Description

Specifications

Specifications

GREM2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GREM2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSRKNRPAG AIPSPYKDGS SNNSERWQHQ IKEVLASSQE ALVVTERKYL KSDWCKTQPL RQTVSEEGCR SRTILNRFCY GQCNSFYIPR HVKKEEESFQ SCAFCKPQRV TSVLVELECP GLDPPFRLKK IQKVKQCRCM SVNLSDSDKQ
Videos
SDS
Documents