missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GSS Protein

A cDNA sequence encoding the GSS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12055-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979218

  • 137.00€ / 5µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GSS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHQH SSGLVPRGSH MATNWGSLLQ DKQQLEELAR QAVDRALAEG VLLRTSQEPT SSEVVSYAPF TLFPSLVPSA LLEQAYAVQM DFNLLVDAVS QNAAFLEQTL SSTIKQDDFT ARLFDIHKQV LKEGIAQTVF LGLNRSDYMF QRSADGSPAL KQIEINTISA SFGGLASRTP AVHRHVLSVL SKTKEAGKIL SNNPSKGLAL GIAKAWELYG SPNALVLLIA QEKERNIFDQRAIENELLAR NIHVIRRTFE DISEKGSLDQ DRRLFVDGQE IAVVYFRDGY MPRQYSLQNW EARLLLERSH AAKCPDIATQ LAGTKKVQQE LSRPGMLEML LPGQPEAVAR LRATFAGLYS LDVGEEGDQA IAEALAAPSR FVLKPQREGG GNNLYGEEMV QALKQLKDSE ERASYILMEK IEPEPFENCL LRPGSPARVV QCISELGIFG VYVRQEKTLV MNKHVGHLLR TKAIEHADGG VAAGVAVLDN PYPV
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The GSS solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions