missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GZMK Protein

A cDNA sequence encoding the GZMK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12073-HIS-EC-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979278

  • 4090.00€ / 1mg
Estimated Shipment:
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3003
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
ADPIIGGKEV SPHSRPFMAS IQYGGHHVCG GVLIDPQWVL TAAHCQYRFT KGQSPTVVLG AHSLSKNEAS KQTLEIKKFI PFSRVTSDPQ SNDIMLVKLQ TAAKLNKHVK MLHIRSKTSL RSGTKCKVTG WGATDPDSLR PSDTLREVTV TVLSRKLCNS QSYYNGDPFI TKDMVCAGDA KGQKDSCKGD SGGPLICKGV FHAIVSGGHE CGVATKPGIY TLLTKKYQTW IKSNLVPPHT NHHHHHH
Greater than 95.0% as determined by analysis by SDS-PAGE.
GZMK Protein
Research Use Only
GZMK
Human
His
GZMK protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions