missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HbA1c Protein
Click to view available options
Quantity:
10 mg
100 μg
500 μg
Unit Size:
100µg
10mg
500µg
Specifications
Specifications
| Name | HbA1c Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human |
| Protein Tag | Untagged |
| Buffer | The HbA1c is supplied in a proprietary buffer formulation having pH 8.0. The HbA1c was dispensed on the basis of HbA1c not total hemoglobin. |
| Purity or Quality Grade | Greater than 96.0%. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction