missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDHD2 Protein
A cDNA sequence encoding the HDHD2 was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Gene ID (Entrez) | 84064 |
---|---|
Name | HDHD2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | HDHD2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989698
|
enQuireBio™
QP12209-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979698
|
enQuireBio™
QP12209-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969698
|
enQuireBio™
QP12209-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
84064 | |
Research Use Only | |
HDHD2 | |
Human | |
His | |
The HDHD2 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT, and 10% glycerol. |
HDHD2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAACRALKAV LVDLSGTLHI EDAAVPGAQE ALKRLRGASV IIRFVTNTTK ESKQDLLERL RKLEFDISED EIFTSLTAAR SLLERKQVRP MLLVDDRALP DFKGIQTSDP NAVVMGLAPE HFHYQILNQA FRLLLDGAPL IAIHKARYYK RKDGLALGPG PFVTALEYAT DTKATVVGKP EKTFFLEALR GTGCEPEEAV MIGDDCRDDV GGAQDVGMLG ILVKTGKYRA SDEEKINPPP YLTCESFPHA VDHILQHLL | |
Greater than 95% as determined by SDS-PAGE. |