missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hemopexin Protein

A cDNA sequence encoding the Hemopexin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12215-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989718

  • 212.00€ / 20µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Hemopexin Protein
Research Use Only
Recombinant Protein
E. coli
MARVLGAPVA LGLWSLCWSL AIATPLPPTS AHGNVAEGET KPDPDVTERC SDGWSFDATT LDDNGTMLFF KGEFVWKSHK WDRELISERW KNFPSPVDAA FRQGHNSVFL IKGDKVWVYP PEKKEKGYPK LLQDEFPGIP SPLDAAVECH RGECQAEGVL FFQGDREWFW DLATGTMKER SWPAVGNCSS ALRWLGRYYC FQGNQFLRFD PVRGEVPPRY PRDVRDYFMP CPGRGHGHRN GTGHGNSTHH GPEYMRCSPH LVLSALTSDN HGATYAFSGT HYWRLDTSRD GWHSWPIAHQ WPQGPSAVDA AFSWEEKLYL VQGTQVYVFL TKGGYTLVSG YPKRLEKEVG TPHGIILDSV DAAFICPGSS RLHIMAGRRL WWLDLKSGAQ ATWTELPWPH EKVDGALCME KSLGPNSCSA NGPGLYLIHG PNLYCYSDVE KLNAAKALPQ PQNVTSLLGC TH
Greater than 95.0%.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Lyophilized from (1.22 mg/ml) solution containing 11.9mM phosphate buffer, 137mM NaCl and 2.7mM KCl, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions