missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSCB Protein
A cDNA sequence encoding the HSCB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12319-1mg
Additional Details : Weight : 0.01000kg
Specifications
HSCB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAASQA GSNYPRCWNC GGPWGPGRED RFFCPQCRAL QAPDPTRDYF SLMDCNRSFR VDTAKLQHRY QQLQRLVHPD FFSQRSQTEK DFSEKHSTLV NDAYKTLLAP LSRGLYLLKL HGIEIPERTD YEMDRQFLIE IMEINEKLAE AESEAAMKEI ESIVKAKQKE FTDNVSSAFE QDDFEEAKEI LTKMRYFSNI EEKIKLKKIP L | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HSCB solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, and 10% glycerol. |