missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-gamma (HEK293-expressed) Protein

Product Code. 18749276
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
This item is not returnable. View return policy

Product Code. 18749276

Brand: R&D Systems™ 10067IF025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Noncovalent homodimer

The Recombinant Human IFN-gamma (HEK293-expressed) Protein is derived from HEK293. The Recombinant Human IFN-gamma (HEK293-expressed) Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000610.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 3458
Molecular Weight (g/mol) 16.8 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human IFN-gamma protein Met1-Gln166
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.