missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF-2 / IGF-II Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_000603.1 |
| Gene ID (Entrez) | 3481 |
| Name | IGF-2 / IGF-II Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | IGF2 |
| Biological Activity | Shows bioactivity at a concentration of 15 ng/mL in the Mouse FDC-P1 cell proliferation assay. The bioactivity acceptance criteria is set at 6.7 x 10^4 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction