missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGFBP 4 Protein
A cDNA sequence encoding the IGFBP 4 was constructed and used to recombinantly synthesize the protein.
137.00€ - 5905.00€
Specifications
Name | IGFBP 4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 range is 0.01-0.09microg/ml as measured by its ability to inhibit the biological activity of IGFII (20ng/ml) on MCF7 human breast adenocarcinoma cells. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934648
|
enQuireBio™
QP10684-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924648
|
enQuireBio™
QP10684-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914648
|
enQuireBio™
QP10684-1MG |
1 mg |
5905.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IGFBP 4 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
DEAIHCPPCS EEKLARCRPP VGCEELVREP GCGCCATCAL GLGMPCGVYT PRCGSGLRCY PPRGVEKPLH TLMHGQGVCM ELAEIEAIQE SLQPSDKDEG DHPNNSFSPC SAHDRRCLQK HFAKIRDRST SGGKMKVNGA PREDARPVPQ GSCQSELHRA LERLAASQSR THEDLYIIPI PNCDRNGNFH PKQCHPALDG QRGKCWCVDR KTGVKLPGGL EPKGELDCHQ LADSFRE + His Tag | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
The ED50 range is 0.01-0.09microg/ml as measured by its ability to inhibit the biological activity of IGFII (20ng/ml) on MCF7 human breast adenocarcinoma cells. | |
Human | |
His | |
IGFBP 4 was lyophilized after extensive dialysis against PBS. |