missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein
Recombinant Protein
3430.00€ - 5600.00€
Specifications
Accession Number | NP_000567.1 |
---|---|
Gene ID (Entrez) | 3553 |
Name | IL-1 beta / IL1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935859
|
enQuireBio™
QP5290-500UG |
500 μg |
3430.00€
500µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925859
|
enQuireBio™
QP5290-1MG |
1 mg |
5600.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_000567.1 | |
IL-1 beta / IL1B Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 12 pg/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 8.3 x 10^7 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
3553 | |
Research Use Only | |
IL1B | |
Recombinant Protein | |
E. coli | |
MAPVRSLNCT LRDSQQKSLV MSGPYELKAL HLQGQDMEQQ VVFSMSFVQG EESNDKIPVA LGLKEKNLYL SCVLKDDKPT LQLESVDPKN YPKKKMEKRF VFNKIEINNK LEFESAQFPN WYISTSQAEN MPVFLGGTKG GQDITDFTMQ FVSS | |
Greater than 95% as determined by SDS-PAGE. |