missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-15 GMP Protein, CF

Product Code. 30306093
Click to view available options
:
1 mg
This item is not returnable. View return policy

Product Code. 30306093

Brand: R&D Systems™ BT015GMP01M/LQ

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. The ED50 for this effect is 0.300-2.60 ng/mL. The specific activity of recombinant human IL-15 is >2.00 x 108 units/mg, which is calibrated against the human IL-15 reference standard (NIBSC code: 95/554).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40933.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3600
Molecular Weight (g/mol) M.W.-theoretical: 13 kDa, M.W.-observed: 9 kDa, under reducing conditions.
Source E. coli-derived human IL-15 protein Asn49-Ser162 Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 6 months when stored at ≤ -65 °C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, ≤ -20 °C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias IL15, IL-15MGC9721, interleukin 15, interleukin-15
Conjugate Unconjugated
Purity or Quality Grade 1 mg
Protein IL-15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.