missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-2 Biotinylated Protein

Product Code. 18723429
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. Quantity unitSize
18723429 25 μg 25µg
1 options
This item is not returnable. View return policy

Product Code. 18723429

Brand: R&D Systems™ BT202025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-2 Biotinylated Protein is derived from E. coli. The Recombinant Human IL-2 Biotinylated Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P60568
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 15 kDa (unlabeled)
Name aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Quantity 25 μg
Source E. coli-derived human IL-2 protein Ala21-Thr153,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 250μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.