missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 23 Protein

Product Code. p-7148513
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15934918

Brand: enQuireBio™ QP107042ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A cDNA sequence encoding the IL 23 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 51561
Name IL 23 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol IL23A
Biological Activity The activity, as determined by the dose dependent secretion of IL-17 in response to IL-23 using murine splenocytes, the ED50 is 1.9ng/ml.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human (HEK293)
Protein Tag Untagged
Sequence MLGSRAVMLL LLLPWTAQGR AVPGGSSPAW TQCQQLSQKL CTLAWSAHPL VGHMDLREEG DEETTNDVPH IQCGDGCDPQ GLRDNSQFCL QRIHQGLIFY EKLLGSDIFT GEPSLLPDSP VGQLHASLLG LSQLLQPEGH HWETQQIPSL SPSQPWQRLL LRFKILRSLQ AFVAVAARVF AHGAATLSP
Buffer IL-23 was lyophilized from a 0.1microm filtered solution (1.05 mg/ml) containing 1xPBS.
Purity or Quality Grade Greater than 95.0% as determined by analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.