missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 23 Protein

Product Code. p-7150257
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15920289

Brand: enQuireBio™ QP123962ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IL 23 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 51561
Name IL 23 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol IL23A
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence IL12B: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS IL23A: RAVPGGSSPA WTQCQQLSQK LCTLAWSAHP LVGHMDLREE GDEETTNDVP HIQCGDGCDP QGLRDNSQFC LQRIHQGLIF YEKLLGSDIF TGEPSLLPDS PVGQLHASLL GLSQLLQPEG HHWETQQIPS LSPSQPWQRL LLRFKILRSL QAFVAVAARV FAHGAATLSP HHHHHH
Buffer IL23 protein solution (1 mg/ml) contains PBS (pH 7.4) and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.