missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17D Protein
A cDNA sequence encoding the IL17D was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Gene ID (Entrez) | 53342 |
---|---|
Name | IL17D Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | IL17D |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934978
|
enQuireBio™
QP10724-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924978
|
enQuireBio™
QP10724-25UG |
25 μg |
212.00€
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914978
|
enQuireBio™
QP10724-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
53342 | |
Research Use Only | |
IL17D | |
Recombinant Protein | |
E. coli | |
APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP | |
Greater than 97.0% as determined by SDS-PAGE. |
IL17D Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Immobilized rHuIL-17D binds to rHuIL-17BR with EC50 less than 2microg/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |