missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL34 Protein

Product Code. p-7148537
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
This item is not returnable. View return policy

Product Code. 15954988

Brand: enQuireBio™ QP107282ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 146433
Name IL34 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol IL34
Biological Activity The specific activity was measured by its ability to induce secretion of MCP-1 from PBMCs.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human (HEK293)
Protein Tag His
Sequence NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP QGLTDVEVSP APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH
Buffer The IL34 was lyophilized from a solution containing 20mM Sodium Citrate, pH 3.0 and 75mM NaCl.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.