missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IMPDH1 Protein

A cDNA sequence encoding the IMPDH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12437-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920419

  • 120.00€ / 2µg

Estimated Shipment: 22-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

3614
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
IMPDH1
Human
His
IMPDH1 Human solution containing 20mM Tris-HCl pH 8.0, 1mM DTT and 20% glycerol.
IMPDH1 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MADYLISGGT GYVPEDGLTA QQLFASADGL TYNDFLILPG FIDFIADEVD LTSALTRKIT LKTPLISSPM DTVTEADMAI AMALMGGIGF IHHNCTPEFQ ANEVRKVKKF EQGFITDPVV LSPSHTVGDV LEAKMRHGFS GIPITETGTM GSKLVGIVTS RDIDFLAEKD HTTLLSEVMT PRIELVVAPA GVTLKEANEI LQRSKKGKLP IVNDCDELVA IIARTDLKKN RDYPLASKDS QKQLLCGAAV GTREDDKYRL DLLTQAGVDV IVLDSSQGNS VYQIAMVHYI KQKYPHLQVI GGNVVTAAQA KNLIDAGVDG LRVGMGCGSI CITQEVMACG RPQGTAVYKV AEYARRFGVP IIADGGIQTV GHVVKALALG ASTVMMGSLL AATTEAPGEY FFSDGVRLKK YRGMGSLDAM EKSSSSQKRY FSEGDKVKIA QGVSGSIQDK GSIQKFVPYL IAGIQHGCQD IGARSLSVLR SMMYSGELKF EKRTMSAQIE GGVHGLHSYE KRLY
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions