missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Integrin alpha V beta 6 Fc Protein, CF

Product Code. 30306764
Click to view available options
:
50 μg
This item is not returnable. View return policy

Product Code. 30306764

Brand: R&D Systems™ 11649AV050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fc Chimera

Measured by its binding ability in a functional ELISA. Recombinant Human Integrin alpha V beta 6 Fc Chimera (Catalog # 11649-AV) binds to Recombinant Human LAP (TGF-β1) (Catalog # 246-LP ) with an ED50 of 9.00-96.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3685
Molecular Weight (g/mol) MolecularWeight-observed: >190 kDa, under non-reducing conditions, MolecularWeight-theroretical: 133 kDa (Integrin alpha V) & 101 kDa (Integrin beta 6)
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Integrin alpha V beta 6 protein Human ITGAV (Phe31-Val992) (N-terminus) Accession # AAA36808.1 IEGR Human IgG1 (Glu99-Lys330) (with modifications) Human ITGB6 (Gly22-Asn707) (N-terminus) Accession # P18564.2 HHPIEGR Human IgG1 (Glu99-Lys330) (with modifications) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Alias CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Protein Integrin alpha V beta 6
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.