missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin-21 / IL-21 Protein
Recombinant Protein
112.00€ - 7420.00€
Specifications
Accession Number | NP_001193935.1 |
---|---|
Gene ID (Entrez) | 59067 |
Name | Interleukin-21 / IL-21 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936039
|
enQuireBio™
QP5394-2UG |
2 μg |
112.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916039
|
enQuireBio™
QP5394-10UG |
10 μg |
263.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906039
|
enQuireBio™
QP5394-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946039
|
enQuireBio™
QP5394-500UG |
500 μg |
4515.00€
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926039
|
enQuireBio™
QP5394-1MG |
1 mg |
7420.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001193935.1 | |
Interleukin-21 / IL-21 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Measured by mino cell proliferation. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
59067 | |
Research Use Only | |
IL21 | |
Recombinant Protein | |
E. coli | |
MLKTLNLGSC VIATNLQEIR NGFSEIRGSV QAKDGNIDIR ILRRTESLQD TKPANRCCLL RHLLRLYLDR VFKNYQTPDH YTLRKISSLA NSFLTIKKDL RLCHAHMTCH CGEEAMKKYS QILSHFEKLE PQAAVVKALG ELDILLQWME ETE | |
Greater than 95% as determined by SDS-PAGE. |