missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Involucrin Protein

A cDNA sequence encoding the Involucrin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12453-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930469

  • 376.00€ / 5µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

3713
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSQQHTLPVT LSPALSQELL KTVPPPVNTH QEQMKQPTPL PPPCQKVPVE LPVEVPSKQE EKHMTAVKGL PEQECEQQQK EPQEQELQQQ HWEQHEEYQK AENPEQQLKQ EKTQRDQQLN KQLEEEKKLL DQQLDQELVK RDEQLGMKKE QLLELPEQQE GHLKHLEQQE GQLKHPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPEQQE GQLELPQQQE GQLELSEQQE GQLELSEQQE GQLKHLEHQE GQLEVPEEQM GQLKYLEQQE GQLKHLDQQE KQPELPEQQM GQLKHLEQQE GQPKHLEQQE GQLEQLEEQE GQLKHLEQQE GQLEHLEHQE GQLGLPEQQV LQLKQLEKQQ GQPKHLEEEE GQLKHLVQQE GQLKHLVQQE GQLEQQERQV EHLEQQVGQL KHLEEQEGQL KHLEQQQGQL EVPEQQVGQP KNLEQEEKQL ELPEQQEGQV KHLEKQEAQL ELPEQQVGQP KHLEQQEKHL EHPEQQDGQL KHLEQQEGQL KDLEQQKGQL EQPVFAPAPG QVQDIQPALP TKGEVLLPVE HQQQKQEVQW PPKHK
Involucrin Protein
Research Use Only
IVL
Human
GST
Involucrin at 100microg/ml in 50mM Tris-HCl, pH 7.5, 10mM L-glutathione (reduced).
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions