missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JDP2 Protein
A cDNA sequence encoding the JDP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12470-5ug
Additional Details : Weight : 0.01000kg
Specifications
JDP2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
JDP2 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 1mM DTT, 1mM EDTA and 250mM imidazole. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMMPGQI PDPSVTTGSL PGLGPLTGLP SSALTVEELK YADIRNLGAM IAPLHFLEVK LGKRPQPVKS ELDEEEERRK RRREKNKVAA ARCRNKKKER TEFLQRESER LELMNAELKT QIEELKQERQ QLILMLNRHR PTCIVRTDSV KTPESEGNPL LEQLEKK |