missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KAT2A Protein

A cDNA sequence encoding the KAT2A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12477-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930549

  • 125.00€ / 5µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

KAT2A Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
KAT2A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 40% glycerol, 200mM NaCl and 1mM EDTA.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGGGSNSSLS LDSAGAEPMP GEKRTLPENL TLEDAKRLRV MGDIPMELVN EVMLTITDPA AMLGPETSLL SANAARDETA RLEERRGIIE FHVIGNSLTP KANRRVLLWL VGLQNVFSHQ LPRMPKEYIA RLVFDPKHKT LALIKDGRVI GGICFRMFPT QGFTEIVFCA VTSNEQVKGY GTHLMNHLKE YHIKHNILYF LTYADEYAIG YFKKQGFSKD IKVPKSRYLG YIKDYEGATL MECELNPRIP YTELSHIIKK QKEIIKKLIE RKQAQIRKVY PGLSCFKEGV RQIPVESVPG IRETGWKPLG KEKGKELKDP DQLYTTLKNL LAQIKSHPSA WPFMEPVKKS EAPDYYEVIR FPIDLKTMTE RLRSRYYVTR KLFVADLQRV IANCREYNPP DSEYCRCASA LEKFFYFKLK EGGLIDK
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions