missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCTD11 Protein
A cDNA sequence encoding the KCTD11 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12480-10ug
Additional Details : Weight : 0.01000kg
Specifications
KCTD11 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLGAMFRAGT PMPPNLNSQG GGHYFIDRDG KAFRHILNFL RLGRLDLPRG YGETALLRAE ADFYQIRPLL DALRELEASQ GTPAPTAALL HADVDVSPRL VHFSARRGPH HYELSSVQVD TFRANLFCTD SECLGALRAR FGVASGDRAE GSPHFHLEWA PRPVELPEVE YGRLGLQPLW TGGPGERREV VGTPSFLEEV LRVALEHGFR LDSVFPDPED LLNSRSLRFV RH | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KCTD11 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |